Learn More
Invitrogen™ SRC1 Recombinant Rabbit Monoclonal Antibody (3S4C6)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542905
Description
Positive test controls include: HeLa, 293T, SGC-7901. Immunogen sequence: VARSSTLPPS NSNMVSTRIN RQQSSDLHSS SHSNSSNSQG SFGCSPGSQI VANVALNQGQ ASSQSSNPSL NLNNSPMEGT GISLAQFMSP RRQVTSGLAT RPRMPNNSFP PNISTLSSPV GMTSSACNNN NRSYSNIPVT SLQGMNEGPN NSVGFSASSP VLRQMSSQNS PSRLNIQPAK AESKDNKEIA SILNEMIQSD NSSSDGKPLD SGLLHNNDRL SDGDSKYSQT SHKLVQLLTT TAEQQLRHAD IDTSCKDVLS CTGTSNSASA NSSGGSCPSS HSSLTERHKI LHRLLQEGSP SDITTLSVEP DKKDSASTSV SVTGQVQGNS SIKLELDASK KKESKDHQLL R.
Clone 1135 recognizes SRC-1, steroid-receptor coactivator-1. SRC-1 is a member of a gene family that functions in the full transcriptional activation of the steroid hormone receptor superfamily and contains intrinsic histone acetyltransferase activity. SRC-1 has a molecular weight of 160 kDa and has a basic helix-loop-helix (bHLH)-PAS domain.
Specifications
| SRC1 | |
| Recombinant Monoclonal | |
| 1.55 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15788 | |
| NCOA1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 400-750 of human KAT13A / SRC1 (Q15788). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Immunoprecipitation, Western Blot | |
| 3S4C6 | |
| Unconjugated | |
| NCOA1 | |
| ASV; bHLHe42; bHLHe74; class E basic helix-loop-helix protein 74; c-SRC; F-SRC-1; Hin-2 protein; KAT13A; mNRC-1; NCOA1; NCoA-1; Nuclear receptor coactivator 1; nuclear receptor coactivator protein 1; p60-Src; PAX3/NCOA1 fusion protein; Protein Hin-2; Renal carcinoma antigen NY-REN-52; RIP160; Src1; SRC-1; SRC-1 (-Q); SRC-a/NCoA-1; Steroid receptor coactivator 1; steroid receptor coactivator-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 8648 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.