missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRC1 Antibody (1135), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NB200-310
This item is not returnable.
View return policy
Description
SRC1 Monoclonal specifically detects SRC1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunoprecipitation.
Specifications
| SRC1 | |
| Monoclonal | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Immunoprecipitation 0.5-2 ug/ml, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
| NCOA1 | |
| Amino acids 477-947 of human SRC-1. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This is specific for SRC-1. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunoprecipitation, ChIP Assay | |
| 1135 | |
| Unconjugated | |
| bHLHe42, BHLHE74, Class E basic helix-loop-helix protein 74, EC 2.3.1.48, Hin-2 protein, KAT13A, MGC129719, NCoA-1, nuclear receptor coactivator 1, PAX3/NCOA1 fusion protein, Protein Hin-2, Renal carcinoma antigen NY-REN-52, RIP160bHLHe74F-SRC-1, SRC-1, SRC1MGC129720, Steroid receptor coactivator 1, steroid receptor coactivator-1 | |
| Mouse | |
| 157 kDa | |
| 0.1 mL | |
| Cancer, Chromatin Research, Transcription Factors and Regulators | |
| 8648 | |
| Human, Mouse, Rat, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction