missing translation for 'onlineSavingsMsg'
Learn More

Src Rabbit anti-Human, Mouse, Rat, Clone: 7G6M9, Novus Biologicals™

Product Code. 18387875
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18387875 20 μg 20µL
18355812 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18387875 Supplier Novus Biologicals Supplier No. NBP31567520UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Src Monoclonal antibody specifically detects Src in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen Src
Applications Western Blot, Immunofluorescence, Immunoprecipitation, KnockDown
Classification Monoclonal
Clone 7G6M9
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias c-src, EC 2.7.10, EC 2.7.10.2, pp60c-src, Rous sarcoma, tyrosine kinase pp60c-src, tyrosine-protein kinase SRC-1, v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog, v-src sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog (avian)
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Src (P12931).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer Stem Cells, Protein Kinase, Signal Transduction, Transcription Factors and Regulators, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 6714
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.