Learn More
Eurofins DiscoverX Src Protein, Active
Description
Src family tyrosine kinases are nonreceptor tyrosine kinases that are functionally linked to some receptor-type tyrosine kinases, G-protein-coupled receptors, cytokine receptors, and cell adhesion molecules/receptors. Src family tyrosine kinases are membrane-associated proteins having Src homology 2 (SH2) and SH3 domains, which are important for protein-protein interaction. The activity of Src family tyrosine kinases is regulated through intramolecular interaction between the SH2 domain and the phosphorylated tyrosine at the C-terminal regulatory site (Tyr-527).
EMBL K03218
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 5% glycerol, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 61.7 kDa |
| Quantity | 250 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Full-length |
| Purity or Quality Grade | ≥69% |
| Protein | Src, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.