missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Src, p Tyr527 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-13245
This item is not returnable.
View return policy
Description
Src Polyclonal antibody specifically detects Src in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Src | |
| Polyclonal | |
| PBS, 1% BSA, 20% Glycerol (pH7) | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| c-src, EC 2.7.10, EC 2.7.10.2, pp60c-src, Rous sarcoma, tyrosine kinase pp60c-src, tyrosine-protein kinase SRC-1, v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog, v-src sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog (avian) | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Src (phospho Tyr527). The exact sequence is proprietary. | |
| 100 μg | |
| Cancer Stem Cells, Phospho Specific, Protein Kinase, Signal Transduction, Transcription Factors and Regulators, Tyrosine Kinases | |
| 6714 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction