missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SRC N1 Recombinant Protein

Product Code. 11509266
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11509266

Brand: Invitrogen™ P2904

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length

SRC is a cytoplasmic tyrosine kinase and proto-oncogene involved in the regulation of embryonic development and cell growth. SRCN1 is a neuronal splice variant of SRC.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P12931
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris with 0.05% NP-40, 0.05 mM EDTA, 100 mM NaCl, 1 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 6714
Molecular Weight (g/mol) 60.6 kDa
Name Human SRC N1
Quantity 10 μg
Regulatory Status For research use only. Not for use in diagnostic procedures
Gene Alias SRCN1
Common Name SRC N1
KinaseFamily SRC
Product Type SRC N1 Recombinant Human Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Baculovirus
Protein Family Kinase and inhibitors
Form Liquid
Protein Subtype Cytoplasmic tyrosine kinase
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.