missing translation for 'onlineSavingsMsg'
Learn More

Src Inhibitor 1, MedChemExpress

Product Code. 16419796
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16419796

Brand: Medchem Express HY101053

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Src Inhibitor 1 is a potent, ATP-competitive and selective dual site Src tyrosine kinase inhibitor with IC50 values of 44 nM for Src and 88nM for Lck.

Src Inhibitor 1 is a potent, ATP-competitive and selective dual site Src tyrosine kinase inhibitor with IC50 values of 44 nM for Src and 88nM for Lck.

Specifications

Chemical Name or Material Src Inhibitor 1
CAS 179248-59-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H19N3O3
Quantity 50 mg
Synonym Src Kinase Inhibitor 1 Src-l1
Solubility Information DMSO : 9.09 mg/mL (24.34 mM; Need ultrasonic)
SMILES COC1=CC2=NC=NC(NC3=CC=C(OC4=CC=CC=C4)C=C3)=C2C=C1OC
Molecular Weight (g/mol) 373.4
Formula Weight 373.4
Percent Purity 99.96%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.