missing translation for 'onlineSavingsMsg'
Learn More

Src Inhibitor 1, MedChemExpress

Product Code. 16480046
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480046 2 mg 2mg
16462686 25 mg 25mg
16443248 10 mg 10mg
16419796 50 mg 50mg
16405977 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16480046 Supplier Medchem Express Supplier No. HY101053

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Src Inhibitor 1 is a potent, ATP-competitive and selective dual site Src tyrosine kinase inhibitor with IC50 values of 44 nM for Src and 88nM for Lck.

Src Inhibitor 1 is a potent, ATP-competitive and selective dual site Src tyrosine kinase inhibitor with IC50 values of 44 nM for Src and 88nM for Lck.

Specifications

Chemical Name or Material Src Inhibitor 1
CAS 179248-59-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H19N3O3
Quantity 2 mg
Synonym Src Kinase Inhibitor 1 Src-l1
Solubility Information DMSO : 9.09 mg/mL (24.34 mM; Need ultrasonic)
SMILES COC1=CC2=NC=NC(NC3=CC=C(OC4=CC=CC=C4)C=C3)=C2C=C1OC
Molecular Weight (g/mol) 373.4
Formula Weight 373.4
Percent Purity 99.96%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.