missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRBD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556505
This item is not returnable.
View return policy
Description
Immunogen sequence: LDQTCIHPES YDIAMRFLSS IGGTLYEVGK PEMQQKINSF LEKEGMEKIA ERLQTTVHTL QVIIDGLSQP ESFDFRTDFD KPDF Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 86%.
SRBD1 gene ontology annotations related to this gene include nucleobase-containing compound metabolic process; translation.
Specifications
| SRBD1 | |
| Polyclonal | |
| Unconjugated | |
| SRBD1 | |
| AI461933; C85414; D530025C17Rik; H_NH0576F01.1; RGD1562788; S1 RNA binding domain 1; S1 RNA-binding domain-containing protein 1; Srbd1; WUGSC:H_NH0576F01.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 301665, 55133, 78586 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q497V5, Q8N5C6 | |
| SRBD1 | |
| Recombinant protein corresponding to Human SRBD1. Recombinant protein control fragment (Product #RP-95975). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction