missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SRA1 Monoclonal Antibody (1D4H8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515287
This item is not returnable.
View return policy
Description
MA5-15287 targets SRA in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15287 immunogen is purified recombinant fragment of SRA expressed in E. Coli.
This gene is involved in transcriptional coactivation by steroid receptors. There is currently data suggesting this gene encodes both a non-coding RNA that functions as part of a ribonucleoprotein complex and a protein coding mRNA. Increased expression of both the transcript and the protein is associated with cancer.
Specifications
| SRA1 | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.03% sodium azide | |
| Q9HD15 | |
| SRA1 | |
| Purified recombinant fragment of SRA expressed in E. Coli. | |
| RUO | |
| 10011 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1D4H8 | |
| Unconjugated | |
| SRA1 | |
| AA959952; LOW QUALITY PROTEIN: steroid receptor RNA activator 1; MGC87674; pp7684; RGD:621148}; Sra; sra1; sra1 {ECO:0000312; SRAP; steroid receptor coactivator; steroid receptor RNA activator 1; steroid receptor RNA activator 1 (complexes with NCOA1); steroid receptor RNA activator protein; Straa1; Strra1; Unknown (protein for MGC:128615) | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction