missing translation for 'onlineSavingsMsg'
Learn More

SR9011 hydrochloride, MedChemExpress

Product Code. 16499767
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499767 5 mg 5mg
16483257 50 mg 50mg
16427216 100 mg 100mg
16420296 2 mg 2mg
16417248 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16499767 Supplier Medchem Express Supplier No. HY16988A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SR9011 hydrochloride is a REV-ERBα/β agonist with IC50s of 790 nM and 560 nM for REV-ERBα and REV-ERBβ, respectively.

SR9011 hydrochloride is a REV-ERBα/β agonist with IC50s of 790 nM and 560 nM for REV-ERBα and REV-ERBβ, respectively.

Specifications

Chemical Name or Material SR9011 hydrochloride
Physical Form Powder
CAS 2070014-94-5
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C23H32Cl2N4O3S
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 32 mg/mL (62.08 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(N1CC(CN(CC2=CC=C(Cl)C=C2)CC3=CC=C([N+]([O-])=O)S3)CC1)NCCCCC.[H]Cl
Molecular Weight (g/mol) 515.5
Formula Weight 515.5
Percent Purity 97.83%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.