missing translation for 'onlineSavingsMsg'
Learn More

SR2211, MedChemExpress

Product Code. 16479777
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16479777 5 mg 5mg
16417258 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16479777 Supplier Medchem Express Supplier No. HY16998
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress SR2211 is a potent, selective synthetic RORγ modulator and functions as an inverse agonist, with a Ki of 105 nM and an IC50 of ∼320 nM.

SR2211 is a potent, selective synthetic RORγ modulator and functions as an inverse agonist, with a Ki of 105 nM and an IC50 of ∼320 nM.

Specifications

Chemical Name or Material SR2211
CAS 1359164-11-6
Color Khaki
Purity Grade Notes Research
Molecular Formula C26H24F7N3O
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (189.58 mM; Need ultrasonic)
SMILES FC1=C(C2=CC=C(CN3CCN(CC4=CC=NC=C4)CC3)C=C2)C=CC(C(C(F)(F)F)(O)C(F)(F)F)=C1
Molecular Weight (g/mol) 527.48
Formula Weight 527.48
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.