missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SR140 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-70714
This item is not returnable.
View return policy
Description
SR140 Polyclonal specifically detects SR140 in Human samples. It is validated for Western Blot.
Specifications
| SR140 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| U2SURP | |
| Synthetic peptides corresponding to SR140(U2-associated SR140 protein) The peptide sequence was selected from the middle region of SR140. Peptide sequence KVAPSKWEAVDESELEAQAVTTSKWELFDQHEESEEEENQNQEEESEDEE. | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| 140 kDa Ser/Arg-rich domain protein, fSAPa, KIAA0332, SR140, U2 snRNP-associated SURP domain containing, U2 snRNP-associated SURP motif-containing protein, U2-associated protein SR140 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 23350 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction