missing translation for 'onlineSavingsMsg'
Learn More

SR10067, MedChemExpress

Product Code. 16437637
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16437637 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16437637 Supplier Medchem Express Supplier No. HY117516
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress SR10067 is a potent, selective and brain penetrant Rev-Erbα/β agonist, with IC50 values are 160 and 170 nM for Rev-Erbβ and Rev-Erbα, respectively. SR10067 has anxiolytic activity.

SR10067 is a potent, selective and brain penetrant Rev-Erbα/β agonist, with IC50 values are 160 and 170 nM for Rev-Erbβ and Rev-Erbα, respectively. SR10067 has anxiolytic activity.

Specifications

Chemical Name or Material SR10067
CAS 1380548-02-6
Color White
Purity Grade Notes Research
Molecular Formula C31H31NO3
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (214.79 mM; Need ultrasonic)
SMILES CC(OC1=CC=C(OCC2CC3=CC=CC=C3CN2C(C4=CC=CC5=C4C=CC=C5)=O)C=C1)(C)C
Molecular Weight (g/mol) 465.58
Formula Weight 465.58
Percent Purity 98.26%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.