missing translation for 'onlineSavingsMsg'
Learn More

SR1001, MedChemExpress

Product Code. 16478367
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478367 5 mg 5mg
16435678 10 mg 10mg
16401847 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16478367 Supplier Medchem Express Supplier No. HY13421
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress SR1001 is a selective RORα and RORγt inverse agonist with Kis 172 and 111 nM, respectively.

SR1001 is a selective RORα and RORγt inverse agonist with Kis 172 and 111 nM, respectively.

Specifications

Chemical Name or Material SR1001
CAS 1335106-03-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H13F6N3O4S2
Quantity 5 mg
Solubility Information DMSO : ≥ 39 mg/mL (81.69 mM)
SMILES CC(NC1=NC(C)=C(S(=O)(NC2=CC=C(C(C(F)(F)F)(O)C(F)(F)F)C=C2)=O)S1)=O
Molecular Weight (g/mol) 477.4
Formula Weight 477.4
Percent Purity 99.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.