missing translation for 'onlineSavingsMsg'
Learn More

SR0987, MedChemExpress

Product Code. 16499896
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499896 50 mg 50mg
16472736 25 mg 25mg
16466097 5 mg 5mg
16424616 100 mg 100mg
16413378 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16499896 Supplier Medchem Express Supplier No. HY101454

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SR0987, a SR1078 analog, is a RORγt agonist, with an EC50 of 800 nM. SR0987 increases IL17 expression while repressing the expression of PD-1.

SR0987, a SR1078 analog, is a RORγt agonist, with an EC50 of 800 nM. SR0987 increases IL17 expression while repressing the expression of PD-1.

Specifications

Chemical Name or Material SR0987
Physical Form Solid
CAS 303126-97-8
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C16H10ClF6NO2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 150 mg/mL (377.17 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(NC1=CC=C(C(C(F)(F)F)(O)C(F)(F)F)C=C1)C2=CC=CC=C2Cl
Molecular Weight (g/mol) 397.7
Formula Weight 397.7
Percent Purity 99.54%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.