Learn More
Invitrogen™ SR-BI Recombinant Rabbit Monoclonal Antibody (SR37-06)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA532001
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
High density lipoproteins (HDLs) play a critical role in cholesterol metabolism and their plasma concentrations are inversely correlated with risk for atherosclerosis. The SR-BI binds HDLs and mediates selective uptake of HDL cholesteryl ester. SR-BI binds HDL with high affinity, is expressed primarily in liver and nonplacental steroidgenic tissues, and mediates selective cholesterol uptake by a distinct mechanism. In mice, it seems that SR-BI plays a key role in determining the levels of plasma lipoprotein cholesterol and the accumulation of cholesterol stores in the adrenal gland.
Specifications
| SR-BI | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| P97943, Q61009, Q8WTV0 | |
| SCARB1 | |
| Synthetic peptide within Human SRB1 aa 81-130. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunohistochemistry, Western Blot | |
| SR37-06 | |
| Unconjugated | |
| SCARB1 | |
| AI120173; CD36; CD36 and LIMPII analogous 1; CD36 antigen (collagen type I receptor thrombospondin receptor)-like 1 (scavanger receptor class B type 1); CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1; CD36 antigen (collagen type I receptor, thrombospondin receptor-like 1); CD36 antigen-like 1; Cd36l1; CD36-related class B scavenger receptor haSR-BI; CLA1; Cla-1; Collagen type I receptor, thrombospondin receptor-like 1; D5Ertd460e; haSR-bI; HDL QTL 1; Hdlq1; HDLQTL6; high density lipoprotein receptor; Hlb398; mSR-BI; Scarb1; scavanger receptor class B type 1; scavenger receptor b1; scavenger receptor class B member 1; scavenger receptor class B type I; scavenger receptor class B type II; scavenger receptor class B type III; scavenger receptor class B, member 1; scavenger receptor class B1; SRB1; SR-B1; SRBI; SR-BI; SR-BII | |
| Rabbit | |
| Protein A | |
| RUO | |
| 20778, 25073, 949 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.