missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SR-BI Recombinant Rabbit Monoclonal Antibody (24GB1395)
Rabbit Recombinant Monoclonal Antibody
£172.00 - £488.00
Specifications
| Antigen | SR-BI |
|---|---|
| Clone | 24GB1395 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30275432
|
Invitrogen™
MA557704 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275413
|
Invitrogen™
MA557705 |
20 μL |
£172.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SR-BI Recombinant Monoclonal Antibody for Western Blot, ICC/IF
High density lipoproteins (HDLs) play a critical role in cholesterol metabolism and their plasma concentrations are inversely correlated with risk for atherosclerosis. The SR-BI binds HDLs and mediates selective uptake of HDL cholesteryl ester. SR-BI binds HDL with high affinity, is expressed primarily in liver and nonplacental steroidgenic tissues, and mediates selective cholesterol uptake by a distinct mechanism. In mice, it seems that SR-BI plays a key role in determining the levels of plasma lipoprotein cholesterol and the accumulation of cholesterol stores in the adrenal gland.Specifications
| SR-BI | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Rat | |
| SCARB1 | |
| AI120173; CD36; CD36 and LIMPII analogous 1; CD36 antigen (collagen type I receptor thrombospondin receptor)-like 1 (scavanger receptor class B type 1); CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1; CD36 antigen (collagen type I receptor, thrombospondin receptor-like 1); CD36 antigen-like 1; Cd36l1; CD36-related class B scavenger receptor haSR-BI; CLA1; Cla-1; Collagen type I receptor, thrombospondin receptor-like 1; D5Ertd460e; haSR-bI; HDL QTL 1; Hdlq1; HDLQTL6; high density lipoprotein receptor; Hlb398; mSR-BI; Scarb1; scavanger receptor class B type 1; scavenger receptor b1; scavenger receptor class B member 1; scavenger receptor class B type I; scavenger receptor class B type II; scavenger receptor class B type III; scavenger receptor class B, member 1; scavenger receptor class B1; SRB1; SR-B1; SRBI; SR-BI; SR-BII | |
| SCARB1 | |
| Primary | |
| Affinity chromatography |
| 24GB1395 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P97943, Q8WTV0 | |
| 25073, 949 | |
| A synthesized peptide derived from human Scavenging Receptor SR-BI (aa 80-110). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title