missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SR-BI Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-54750-25ul
This item is not returnable.
View return policy
Description
SR-BI Polyclonal specifically detects SR-BI in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| SR-BI/SR-BII | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL | |
| CD36 and LIMPII analogous 1, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1, CD36L1scavenger receptor class B type 1, CLA1CD36 antigen-like 1, CLA-1HDLQTL6, Collagen type I receptor, thrombospondin receptor-like 1, scavenger receptor class B type III, scavenger receptor class B, member 1, SRB1scavenger receptor class B member 1, SR-BIMGC138242 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| SCARB1 | |
| This SR-BI Antibody was developed against a Recombinant Protein corresponding to amino acids: PRSLSFNMWKEIPIPFYLSVYFFDVMNPSEILKGEKPQVRERGPYVYREFRHKSNITFNNNDTVSFLEYRTFQFQPSKSHGSESDYIVMPNILVLGA | |
| 25 μL | |
| Breast Cancer, Cancer, Cardiovascular Biology, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction, Virology Bacteria and Parasites | |
| 949 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction