missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SR-BI Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB400-104AF594
This item is not returnable.
View return policy
Description
SR-BI Polyclonal specifically detects SR-BI in Human, Mouse, Rat, Porcine, Chinese Hamster, Hamster, Mustelid, Primate, Rabbit, Golden Syrian Hamster samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated, Knockdown Validated.
Specifications
| SR-BI | |
| Polyclonal | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated, Knockdown Validated | |
| CD36 and LIMPII analogous 1, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1, CD36L1scavenger receptor class B type 1, CLA1 CD36 antigen-like 1, CLA-1 HDLQTL6, Collagen type I receptor, thrombospondin receptor-like 1, MGC138242, SCARB1, scavenger receptor class B type III, scavenger receptor class B, member 1, SR-B1, SRB1 scavenger receptor class B member 1, SRBI | |
| Rabbit | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Pig, Chinese Hamster, Hamster, Mustelid, Primate, Rabbit, Golden Syrian Hamster | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay, KnockDown | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SCARB1 | |
| A C-terminal peptide containing residues from mouse SR-BI (within residues 450-509). [UniProt Q61009] | |
| Breast Cancer, Cancer, Cardiovascular Biology, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction, Virology Bacteria and Parasites | |
| 949 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction