missing translation for 'onlineSavingsMsg'
Learn More

SR-BI Antibody (185-1G2 (B467)), PE, Novus Biologicals™

Product Code. 18350223 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18350223 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18350223 Supplier Novus Biologicals Supplier No. NBP312073PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SR-BI Monoclonal antibody specifically detects SR-BI in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen SR-BI
Applications Flow Cytometry
Classification Monoclonal
Clone 185-1G2 (B467)
Conjugate PE
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD36 and LIMPII analogous 1, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1, CD36L1scavenger receptor class B type 1, CLA1 CD36 antigen-like 1, CLA-1 HDLQTL6, Collagen type I receptor, thrombospondin receptor-like 1, MGC138242, SCARB1, scavenger receptor class B type III, scavenger receptor class B, member 1, SR-B1, SRB1 scavenger receptor class B member 1, SRBI
Host Species Rabbit
Immunogen This antibody was raised by immunizing mice with stimulated human leukocytes.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cardiovascular Biology, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 949
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.