missing translation for 'onlineSavingsMsg'
Learn More

SR 95531 hydrobromide, Tocris Bioscience™

Product Code. 30001219
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Competitive and selective GABAA antagonist

SR 95531 hydrobromide is a selective, competitive GABAA receptor antagonist and allosteric inhibitor (IC50 = 200 nM). SR 95531 displaces [3H]-GABA from rat brain membranes with a Ki of 150 nM. Unlike Bicuculline (Cat. No. 0130), SR 95531 selectively antagonizes GABA-induced Cl- currents with little action on Pentobarbitone -induced currents (Cat. No. 4579). SR 95531 can act as an agonist at high concentrations (>100 μM) and is a weak agonist at GABAA receptors with α1β2(Y157S)γ2L subunits. SR 95531 also acts as a competitive antagonist of recombinant glycine receptors and exhibits greater potency at glycine receptors containing β subunits.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104104-50-9
Molecular Formula C15H18BrN3O3
Molecular Weight (g/mol) 368.231
InChI Key GFZHNFOGCMEYTA-UHFFFAOYSA-N
Synonym gabazine, sr 95531 hydrobromide, gabazine hydrobromide,sr-95531, 1 6h-pyridazinebutanoic acid, 6-imino-3-4-methoxyphenyl-, monohydrobromide, 4-6-imino-3-4-methoxyphenyl pyridazin-1 6h-yl butanoic acid hydrobromide, 6-amino-5-methyl-3-4-methoxyphenyl-1-pyridaziniumbutyric acid br, 6-imino-3-4-methoxyphenyl-1 6h-pyridazinebutanoic acid hydrobromide, 6-imino-3-4-methoxyphenyl-1 6h-pyridazinebutanoic acid monohydrobromide, 1 6h-pyridazinebutanoicacid, 6-imino-3-4-methoxyphenyl-, hydrobromide 1:1, gabazine hydrobromide
PubChem CID 107895
IUPAC Name 4-[6-imino-3-(4-methoxyphenyl)pyridazin-1-yl]butanoic acid;hydrobromide
SMILES COC1=CC=C(C=C1)C2=NN(C(=N)C=C2)CCCC(=O)O.Br

Specifications

Quantity 10 mg
Formula Weight 368.23
Percent Purity >98%
Chemical Name or Material SR 95531 hydrobromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.