missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ SR 9243

Product Code. 18797536
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18797536 10 mg 10mg
18746596 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18797536 Supplier R&D Systems™ Supplier No. 6138/10
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective LXR inverse agonist

SR 9243 is a selective LXR inverse agonist. Exhibits no significant activity at other nuclear receptors at maximally effective concentration. Inhibits the Warburg effect and lipogenesis by down-regulation of LXR-mediated gene expression. Selectively reduces cell viability and induces apoptosis in cancer cell lines in vitro. Suppresses hepatic steatosis and inhibits growth of tumor xenografts in mice.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1613028-81-1
Quantity 10 mg
Solubility Soluble to 20mM in DMSO
Product Line Tocris
Molecular Formula C31H32BrNO4S2
Formula Weight 626.62g/mol
Color White
Product Type Inhibitor

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.