missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SR-4370, MedChemExpress
Description
SR-4370 is an inhibitor of HDAC, with IC50s of 0.13 μM, 0.58 μM, 0.006 μM, 2.3 μM, and 3.4 μM for HDAC1, HDAC2, HDAC3, HDAC8, and HDAC6, respectively.
Specifications
Specifications
| Chemical Name or Material | SR-4370 |
| CAS | 1816294-67-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C17H18F2N2O |
| Quantity | 100 mg |
| Solubility Information | DMSO : ≥ 150 mg/mL (492.89 mM) |
| SMILES | O=C(C1=CC=C(C2=CC=CC(F)=C2F)C=C1)NNCCCC |
| Molecular Weight (g/mol) | 304.33 |
| Formula Weight | 304.33 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?