missing translation for 'onlineSavingsMsg'
Learn More

SR-3677, MedChemExpress

Product Code. 16491797
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491797 50 mg 50mg
16476156 100 mg 100mg
16415638 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16491797 Supplier Medchem Express Supplier No. HY13300

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SR-3677 is a potent and selective ROCK-II inhibitor with an IC50 of ∼3 nM.

SR-3677 is a potent and selective ROCK-II inhibitor with an IC50 of ∼3 nM.

Specifications

Chemical Name or Material SR-3677
CAS 1072959-67-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H24N4O4
Quantity 50 mg
Solubility Information DMSO : ≥ 43 mg/mL (105.28 mM)
SMILES CN(C)CCOC1=C(NC(C2OC(C=CC=C3)=C3OC2)=O)C=CC(C4=CNN=C4)=C1
Molecular Weight (g/mol) 408.45
Formula Weight 408.45
Percent Purity 99.9%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.