missing translation for 'onlineSavingsMsg'
Learn More

SR-3306, MedChemExpress

Product Code. 16455378
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16455378 10 mg 10mg
16448097 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16455378 Supplier Medchem Express Supplier No. HY12829

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SR-3306 is a selective, potent, highly brain penetrant JNK inhibitor.

SR-3306 is a selective, potent, highly brain penetrant JNK inhibitor.

Specifications

Chemical Name or Material SR-3306
CAS 1128096-91-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C28H26N8O
Quantity 10 mg
Solubility Information DMSO : 125 mg/mL (254.81 mM; Need ultrasonic)
SMILES CC(C=C1)=NC=C1C2=NN(C=N2)C3=CC=C(NC4=NC=CC(C5=CC=CC(N6CCOCC6)=C5)=N4)C=C3
Molecular Weight (g/mol) 490.56
Formula Weight 490.56
Percent Purity 95.11%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.