missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SR 27897
GREENER_CHOICE

Product Code. p-5546037
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18727227

Brand: Tocris Bioscience™ 2190/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective CCK1 antagonist

SR 27897 is a potent, non-peptide CCK1 receptor antagonist that displays > 33-fold selectivity over CCK2 receptors (EC50 values are 6 and 200 nM respectively). Causes an increase in plasma leptin levels and increases food intake in rats in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type SR 27897
Purity >99%
Molecular Weight (g/mol) 411.86
Inhibitors SR 27897

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.