missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SR 27897

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
SR 27897 is a potent, non-peptide CCK1 receptor antagonist that displays > 33-fold selectivity over CCK2 receptors (EC50 values are 6 and 200 nM respectively). Causes an increase in plasma leptin levels and increases food intake in rats in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | SR 27897 |
| Purity | >99% |
| Molecular Weight (g/mol) | 411.86 |
| Inhibitors | SR 27897 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction