missing translation for 'onlineSavingsMsg'
Learn More

SR 202, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18055382
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18055382

Brand: Tocris Bioscience 2022/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective PPARγ antagonist; antidiabetic and antiobesity agent

Selective PPARγ antagonist; antidiabetic and antiobesity agent. Attenuates troglitazone-induced PPARγ transcriptional activity (IC50 = 140 μM) without affecting ligand-stimulated PPARα, PPARβ or FXR transcriptional activity. Inhibits PPARγ-dependent adipocyte differentiation and growth in vitro and in vivo. Improves insulin sensitivity in diabetic ob/ob mice and increases HDL levels in rats in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76541-72-5
Molecular Formula C11H17ClO7P2
Molecular Weight (g/mol) 358.648
InChI Key VQHUQHAPWMNBLP-UHFFFAOYSA-N
Synonym mifobate, clenicor, mifobatum latin, mifobato spanish, mifobate usan:inn, 4-chlorophenyl dimethoxyphosphinyl methyl phosphoric acid dimethyl ester, dimethyl alpha-dimethoxyphosphinyl-p-chlorobenzyl phosphate, dimethyl p-chloro-alpha-hydroxybenzyl phosphonate, dimethyl phosphate, phosphoric acid, 4-chlorophenyl dimethoxyphosphinyl methyl dimethyl ester, dsstox_cid_25741
PubChem CID 60910
IUPAC Name [(4-chlorophenyl)-dimethoxyphosphorylmethyl] dimethyl phosphate
SMILES COP(=O)(C(C1=CC=C(C=C1)Cl)OP(=O)(OC)OC)OC

Specifications

Quantity 10 mg
Formula Weight 358.65
Percent Purity >99%
Chemical Name or Material SR 202
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.