missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SR 142948, Tocris Bioscience™
Highly potent NTS receptor antagonist
£149.02 - £273.35
Chemical Identifiers
CAS | 184162-64-9 |
---|---|
Molecular Formula | C39H51N5O6 |
Molecular Weight (g/mol) | 685.866 |
InChI Key | LWULHXVBLMWCHO-UHFFFAOYSA-N |
Synonym | d0oj7s, sr 142948 hydrochloride, 2-5-2,6-dimethoxyphenyl-1-4-3-dimethylamino propyl methylamino carbonyl-2-1-methylethyl phenyl-1h-pyrazol-3-yl carbonyl amino-tricyclo 3.3.1.13,7 decane-2-carboxylic acid, 2-5-2,6-dimethoxyphenyl-1-4-n-3-dimethylaminopropyl-n-methylcarbamoyl-2-isopropylphenyl-1h-pyrazole3-carbonyl amino adamantane-2-carboxylic acid hydrochloride, 2-5-2,6-dimethoxyphenyl-1-4-3-dimethylaminopropyl-methylcarbamoyl-2-propan-2-ylphenyl pyrazole-3-carbonyl amino adamantane-2-carboxylic acid, 2-5-2,6-dimethoxyphenyl-1-4-3-dimethylamino propyl-methylcarbamoyl-2-propan-2-ylphenyl pyrazole-3-carbonyl amino adamantane-2-carboxylic acid, 2-5-2,6-dimethoxyphenyl-1-4-3-dimethylamino propyl methyl carbamoyl-2-isopropylphenyl pyrazole-3-amido adamantane-2-carboxylic acid, 2-5-2,6-dimethoxyphenyl-1-4-n-methyl-n-3-dimethylaminopropyl carbamoyl-2-isopropylphenyl-3-pyrazolylcarbonylamino-2-adamantanecarboxylic acid |
PubChem CID | 5311451 |
IUPAC Name | 2-[[5-(2,6-dimethoxyphenyl)-1-[4-[3-(dimethylamino)propyl-methylcarbamoyl]-2-propan-2-ylphenyl]pyrazole-3-carbonyl]amino]adamantane-2-carboxylic acid |
SMILES | CC(C)C1=C(C=CC(=C1)C(=O)N(C)CCCN(C)C)N2C(=CC(=N2)C(=O)NC3(C4CC5CC(C4)CC3C5)C(=O)O)C6=C(C=CC=C6OC)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30001411
|
Tocris Bioscience
2309/1 |
1 mg |
£149.02
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18073792
|
Tocris Bioscience
2309/10 |
10 mg |
£273.35
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent non-peptide neurotensin (NT) receptor antagonist that binds with high affinity (IC50 = 0.32 - 3.96 nM; Ki < 10 nM). Attenuates amphetamine-induced hyperactivity and is orally active in vivo.Chemical Identifiers
184162-64-9 | |
685.866 | |
d0oj7s, sr 142948 hydrochloride, 2-5-2,6-dimethoxyphenyl-1-4-3-dimethylamino propyl methylamino carbonyl-2-1-methylethyl phenyl-1h-pyrazol-3-yl carbonyl amino-tricyclo 3.3.1.13,7 decane-2-carboxylic acid, 2-5-2,6-dimethoxyphenyl-1-4-n-3-dimethylaminopropyl-n-methylcarbamoyl-2-isopropylphenyl-1h-pyrazole3-carbonyl amino adamantane-2-carboxylic acid hydrochloride, 2-5-2,6-dimethoxyphenyl-1-4-3-dimethylaminopropyl-methylcarbamoyl-2-propan-2-ylphenyl pyrazole-3-carbonyl amino adamantane-2-carboxylic acid, 2-5-2,6-dimethoxyphenyl-1-4-3-dimethylamino propyl-methylcarbamoyl-2-propan-2-ylphenyl pyrazole-3-carbonyl amino adamantane-2-carboxylic acid, 2-5-2,6-dimethoxyphenyl-1-4-3-dimethylamino propyl methyl carbamoyl-2-isopropylphenyl pyrazole-3-amido adamantane-2-carboxylic acid, 2-5-2,6-dimethoxyphenyl-1-4-n-methyl-n-3-dimethylaminopropyl carbamoyl-2-isopropylphenyl-3-pyrazolylcarbonylamino-2-adamantanecarboxylic acid | |
2-[[5-(2,6-dimethoxyphenyl)-1-[4-[3-(dimethylamino)propyl-methylcarbamoyl]-2-propan-2-ylphenyl]pyrazole-3-carbonyl]amino]adamantane-2-carboxylic acid |
C39H51N5O6 | |
LWULHXVBLMWCHO-UHFFFAOYSA-N | |
5311451 | |
CC(C)C1=C(C=CC(=C1)C(=O)N(C)CCCN(C)C)N2C(=CC(=N2)C(=O)NC3(C4CC5CC(C4)CC3C5)C(=O)O)C6=C(C=CC=C6OC)OC |
Specifications
184162-64-9 | |
d0oj7s, sr 142948 hydrochloride, 2-5-2,6-dimethoxyphenyl-1-4-3-dimethylamino propyl methylamino carbonyl-2-1-methylethyl phenyl-1h-pyrazol-3-yl carbonyl amino-tricyclo 3.3.1.13,7 decane-2-carboxylic acid, 2-5-2,6-dimethoxyphenyl-1-4-n-3-dimethylaminopropyl-n-methylcarbamoyl-2-isopropylphenyl-1h-pyrazole3-carbonyl amino adamantane-2-carboxylic acid hydrochloride, 2-5-2,6-dimethoxyphenyl-1-4-3-dimethylaminopropyl-methylcarbamoyl-2-propan-2-ylphenyl pyrazole-3-carbonyl amino adamantane-2-carboxylic acid, 2-5-2,6-dimethoxyphenyl-1-4-3-dimethylamino propyl-methylcarbamoyl-2-propan-2-ylphenyl pyrazole-3-carbonyl amino adamantane-2-carboxylic acid, 2-5-2,6-dimethoxyphenyl-1-4-3-dimethylamino propyl methyl carbamoyl-2-isopropylphenyl pyrazole-3-amido adamantane-2-carboxylic acid, 2-5-2,6-dimethoxyphenyl-1-4-n-methyl-n-3-dimethylaminopropyl carbamoyl-2-isopropylphenyl-3-pyrazolylcarbonylamino-2-adamantanecarboxylic acid | |
CC(C)C1=C(C=CC(=C1)C(=O)N(C)CCCN(C)C)N2C(=CC(=N2)C(=O)NC3(C4CC5CC(C4)CC3C5)C(=O)O)C6=C(C=CC=C6OC)OC | |
685.866 | |
685.86 | |
SR 142948 |
C39H51N5O6 | |
LWULHXVBLMWCHO-UHFFFAOYSA-N | |
2-[[5-(2,6-dimethoxyphenyl)-1-[4-[3-(dimethylamino)propyl-methylcarbamoyl]-2-propan-2-ylphenyl]pyrazole-3-carbonyl]amino]adamantane-2-carboxylic acid | |
5311451 | |
>97% |
Safety and Handling
Recommended Storage : Store at +4°C