missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SR 140333
GREENER_CHOICE

Product Code. 18716937
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 18716937

Brand: Tocris Bioscience™ 4012/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent NK1 receptor antagonist

Potent NK1 receptor antagonist (Ki = 0.74 nM. IC50 = 1.6 nM). Inhibits substance P -invoked calcium mobilization and outward current (IC50 = 1.3 nM); blocks NK1-mediated nitric oxide-dependent vasodilation in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153050-21-6
Molecular Formula C37H45Cl3N2O2

Specifications

Quantity 10 mg
Formula Weight 656.12
Purity >97%
Molecular Weight (g/mol) 656.129
Chemical Name or Material SR 140333
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.