missing translation for 'onlineSavingsMsg'
Learn More

Squaric Acid Dibutyl Ester, 97%

Product Code. 10368653 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10368653

Brand: Thermo Scientific Acros 133170250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2892-62-8 | C12H18O4 | 226.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2892-62-8
Molecular Formula C12H18O4
Molecular Weight (g/mol) 226.27
MDL Number MFCD00037150
InChI Key XBRWELTXMQSEIN-UHFFFAOYSA-N
Synonym dibutyl squarate, 3,4-dibutoxy-3-cyclobutene-1,2-dione, squaric acid dibutyl ester, sadbe, squaric acid dibutylester, 3,4-di-n-butoxy-3-cyclobutene-1,2-dione, unii-4rto57vg65, 1,2-dibutyl squarate, 3-cyclobutene-1,2-dione, 3,4-dibutoxy, acmc-209h5p
PubChem CID 65108
ChEBI CHEBI:53612
IUPAC Name 3,4-dibutoxycyclobut-3-ene-1,2-dione
SMILES CCCCOC1=C(OCCCC)C(=O)C1=O

Specifications

Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 121.0°C to 122.0°C (0.2 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4930 to 1.4950
Quantity 25 g
Merck Index 15, 8897
Specific Gravity 1.05
Solubility Information Solubility in water: insoluble.
Formula Weight 226.27
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Squaric acid dibutyl ester
Show More Show Less
compliance-icons
Product Identifier
  • Squaric acid dibutyl ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.