missing translation for 'onlineSavingsMsg'
Learn More

Squalene, MedChemExpress

Product Code. p-200096240
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16479208

Brand: Medchem Express HYN1214

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Squalene is an intermediate product in the synthesis of cholesterol, and shows several pharmacological properties such as hypolipidemic, hepatoprotective, cardioprotective, antioxidant, and antitoxicant activity. Squalene also has anti-fungal activity and can be used for the research of Trichophyton mentagrophytes treatment.

Squalene is an intermediate product in the synthesis of cholesterol, and shows several pharmacological properties such as hypolipidemic, hepatoprotective, cardioprotective, antioxidant, and antitoxicant activity. Squalene also has anti-fungal activity and can be used for the research of Trichophyton mentagrophytes treatment.

Specifications

Chemical Name or Material Squalene
Physical Form Liquid
CAS 111-02-4
Color Colorless
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C30H50
For Use With (Application) COVID-19-immunoregulation
Synonym Super Squalene trans-Squalene AddaVax
Solubility Information DMSO : 16.67 mg/mL (40.59 mM; Need ultrasonic)
SMILES C/C(C)=C\CC/C(C)=C/CC/C(C)=C/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)\C
Molecular Weight (g/mol) 410.72
Formula Weight 410.72
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.