missing translation for 'onlineSavingsMsg'
Learn More

Squalene, MedChemExpress

Product Code. 16411628
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16411628 5 mg 5mg
16479208 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16411628 Supplier Medchem Express Supplier No. HYN1214
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Squalene is an intermediate product in the synthesis of cholesterol, and shows several pharmacological properties such as hypolipidemic, hepatoprotective, cardioprotective, antioxidant, and antitoxicant activity. Squalene also has anti-fungal activity and can be used for the research of Trichophyton mentagrophytes treatment.

Squalene is an intermediate product in the synthesis of cholesterol, and shows several pharmacological properties such as hypolipidemic, hepatoprotective, cardioprotective, antioxidant, and antitoxicant activity. Squalene also has anti-fungal activity and can be used for the research of Trichophyton mentagrophytes treatment.

Specifications

Chemical Name or Material Squalene
Physical Form Liquid
CAS 111-02-4
Color Colorless
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C30H50
For Use With (Application) COVID-19-immunoregulation
Synonym Super Squalene trans-Squalene AddaVax
Solubility Information DMSO : 16.67 mg/mL (40.59 mM; Need ultrasonic)
SMILES C/C(C)=C\CC/C(C)=C/CC/C(C)=C/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)\C
Molecular Weight (g/mol) 410.72
Formula Weight 410.72
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.