missing translation for 'onlineSavingsMsg'
Learn More

Squalene, 98%, Thermo Scientific Chemicals

Product Code. p-7025466
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11400007

Brand: Thermo Scientific Alfa Aesar B20944.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Biosynthetic precursor to all steroids Squalene is used as a precursor to steroids and ergosterol and as an immunologic adjuvant in vaccines. It plays a vital role in the preparation of cholesterol, steroid hormones and vitamin D. It serves as a lubricant for fatty acid surfactant films adsorbed on iron oxide surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biosynthetic precursor to all steroids Squalene is used as a precursor to steroids and ergosterol and as an immunologic adjuvant in vaccines. It plays a vital role in the preparation of cholesterol, steroid hormones and vitamin D. It serves as a lubricant for fatty acid surfactant films adsorbed on iron oxide surfaces.

Solubility
Miscible with diethyl ether and acetone. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-02-4
Molecular Formula C30H50
Molecular Weight (g/mol) 410.73
MDL Number MFCD00008912
InChI Key YYGNTYWPHWGJRM-AAJYLUCBSA-N
Synonym squalene, trans-squalene, spinacene, supraene, all-trans-squalene, 2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene, e,e,e,e-squalene, unii-7qwm220fjh, 7qwm220fjh, spinacen
PubChem CID 638072
ChEBI CHEBI:15440
IUPAC Name (6E,10E,14E,18E)-2,6,10,15,19,23-hexamethyltetracosa-2,6,10,14,18,22-hexaene
SMILES CC(C)=CCC\C(C)=C\CC\C(C)=C\CC\C=C(/C)CC\C=C(/C)CCC=C(C)C

Specifications

Melting Point °C to 75°C
Density 0.858
Boiling Point 285°C (25 mm)
Flash Point 218°C (424°F)
Assay Percent Range 0.97
Refractive Index 1.4955
Quantity 100 g
Sensitivity Air and light sensitive
Merck Index 14,8768
Solubility Information Miscible with diethyl ether and acetone. Immiscible with water.
Formula Weight 410.73
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Squalene
Show More Show Less
compliance-icons
Product Identifier
  • Squalene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contains : Squalene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.