missing translation for 'onlineSavingsMsg'
Learn More

Squalane, 98%, Thermo Scientific Chemicals

Product Code. p-7023910
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 11439056

Brand: Thermo Scientific Alfa Aesar A17931.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Squalane influence on the vital pathway that transforms glucose into cholesterol may be pivotal. It regulates or controls the rate of synthesis of the enzyme HMG Co-A reductase, and may contribute to effective treatments for cancer and heart disease. It plays an important role as a terminator in the biomembrane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-01-3
Molecular Formula C30H62
Molecular Weight (g/mol) 422.826
MDL Number MFCD00008953
InChI Key PRAKJMSDJKAYCZ-UHFFFAOYSA-N
Synonym squalane, perhydrosqualene, dodecahydrosqualene, cosbiol, vitabiosol, spinacane, robane, hexamethyltetracosane, tetracosane, 2,6,10,15,19,23-hexamethyl, hexamethyl tetracosane
PubChem CID 8089
IUPAC Name 2,6,10,15,19,23-hexamethyltetracosane
SMILES CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C

Specifications

Melting Point -38°C
Density 0.81
Boiling Point 210°C to 215°C (1 mmHg)
Flash Point 218°C (424°F)
Refractive Index 1.451
Quantity 100 g
Beilstein 776019
Merck Index 14,8767
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 422.83
Percent Purity 98%
Chemical Name or Material Squalane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.