missing translation for 'onlineSavingsMsg'
Learn More

Squalamine lactate, MedChemExpress

Product Code. 16409607
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16409607

Brand: Medchem Express HY16467

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Squalamine lactate is an aminosterol compound discovered in the tissues of the dogfish shark, with antimicrobial activity, and used for the treatment of neovascular age-related macular degeneration.

Squalamine lactate is an aminosterol compound discovered in the tissues of the dogfish shark, with antimicrobial activity, and used for the treatment of neovascular age-related macular degeneration.

Specifications

Chemical Name or Material Squalamine lactate
Physical Form Solid
CAS 320725-47-1
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C37H71N3O8S
For Use With (Application) COVID-19-immunoregulation
Synonym MSI-1256F
Solubility Information DMSO : ≥ 36.66 mg/mL (51.06 mM)
SMILES NCCCCNCCCN[C@H]1CC[C@@]2(C)[C@](C[C@@H](O)[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@H](C)CC[C@@H](OS(=O)(O)=O)C(C)C)([H])C1.C[C@@H](C(O)=O)O
Molecular Weight (g/mol) 718.04
Formula Weight 718.04
Percent Purity 98.37%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.