Learn More
Invitrogen™ SQLE Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5104313
Product Code. 16640765
  • £442.00 / 100µL
Description
Antibody detects endogenous levels of total SQLE.
SQLE, also named as ERG1, SE and SM, belongs to the squalene monooxygenase family. It catalyzes the first oxygenation step in cholesterol synthesis, acting on squalene before cyclization into the basic steroid structure. SQLE may serve as a flux-controlling enzyme beyond 3-hydroxy-3-methylglutaryl-coenzyme A reductase (HMGR, considered as rate limiting). It is also post translationally regulated by cholesterol-dependent proteasomal degradation. The MW of SQLE is about 64 kDa.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SQLE
Polyclonal
Unconjugated
SQLE
AI323792; Erg1; FLJ30795; SE; Sqle; Squalene epoxidase; squalene monooxygenase; Squalene monooxygenase-like protein
Rabbit
Affinity chromatography
RUO
20775, 29230, 6713
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P52019, P52020, Q14534
SQLE
A synthesized peptide derived from human SQLE(Accession Q14534), corresponding to amino acid residues W456-V506.
100 μL
Primary
Human, Mouse, Rat