missing translation for 'onlineSavingsMsg'
Learn More

SQ22536, MedChemExpress

Product Code. 16475727
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475727 5 mg 5mg
16490906 200 mg 200mg
16469556 50 mg 50mg
16462988 10 mg 10mg
16422576 25 mg 25mg
16414306 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16475727 Supplier Medchem Express Supplier No. HY100396
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress SQ22536 is an effective adenylate cyclase (AC) inhibitor.

SQ22536 is an effective adenylate cyclase (AC) inhibitor.

Specifications

Chemical Name or Material SQ22536
CAS 17318-31-9
Color White
Purity Grade Notes Research
Molecular Formula C9H11N5O
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (487.28 mM; Need ultrasonic) ∣H2O : 55 mg/mL (268.01 mM; Need ultrasonic)
SMILES NC1=C2N=CN(C3OCCC3)C2=NC=N1
Molecular Weight (g/mol) 205.22
Formula Weight 205.22
Percent Purity 98.41%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.