missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPZ1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA523391
This item is not returnable.
View return policy
Description
The amino acid sequence used as immunogen is 82% homologous in Chicken.
Spermatogenic leucine zipper protein 1 is a member of the basic helix-loop-helix (bHLH) family of transcription factors which is expressed specifically in the testis and epididymis. Studies show that SPZ1 acts downstream of mitogen-activated protein kinase (MAPK), an extracellular signal-regulated kinase 1/2 to up-regulate cell proliferation and tumorigenesis. Also, through an interaction with proliferating cell nuclear antigen (PCNA) promoter, SPZ1 induced cell proliferation concomitant with an increase in PCNA gene expression.
Specifications
| SPZ1 | |
| Polyclonal | |
| Unconjugated | |
| Spz1 | |
| 1700027M20Rik; AI429130; bhlh-zip transcription factor SPZ1; NYD-TSP1; PPP1R148; protein phosphatase 1, regulatory subunit 148; spermatogenic leucine zipper 1; spermatogenic leucine zipper protein 1; Spermatogenic Zip 1; SPZ1; testis secretory sperm-binding protein Li 232m; testis-specific protein 1; Testis-specific protein NYD-TSP1; TSP1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 499510, 79401, 84654 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.05% BSA and 0.05% sodium azide | |
| Q6AXY9, Q99MY0, Q9BXG8 | |
| Spz1 | |
| A portion of amino acids 40-90 of human SPZ1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction