missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPTLC3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-54899-25ul
This item is not returnable.
View return policy
Description
SPTLC3 Polyclonal specifically detects SPTLC3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| SPTLC3 | |
| Polyclonal | |
| Unconjugated | |
| 40% glycerol and PBS (pH 7.2) with 0.02% Sodium Azide | |
| SPTLC3 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence: MANPGGGAVCNGKLHNHKKQSNGSQSRNCTKNGIVKEAQQNGKPHFYDKLIVESFEEA | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.1 mg/mL | |
| Western Blot 0.4 μg/mL, Immunocytochemistry/Immunofluorescence | |
| C20orf38, Chromosome 20 Open Reading Frame 38, dJ718P11, EC 2.3.1.50, hLCB2b, LCB 3, LCB2B, LCB3, Long Chain Base Biosynthesis Protein 2b, Long Chain Base Biosynthesis Protein 3, Serine Palmitoyltransferase 3, Serine Palmitoyltransferase, Long Chain Base Subunit 2-Like (Aminotransferase 2), Serine Palmitoyltransferase, Long Chain Base Subunit 3, Serine-Palmitoyl-CoA Transferase 3, SPT 3, SPT3, SPTLC2L | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 55304 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction