missing translation for 'onlineSavingsMsg'
Learn More

SPTLC3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-54899

 View more versions of this product

Product Code. 18212180

  • £513.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SPTLC3 Polyclonal specifically detects SPTLC3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SPTLC3
Polyclonal
Unconjugated
40% glycerol and PBS (pH 7.2) with 0.02% Sodium Azide
SPTLC3
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence: MANPGGGAVCNGKLHNHKKQSNGSQSRNCTKNGIVKEAQQNGKPHFYDKLIVESFEEA
100 μL
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot, Immunocytochemistry, Immunofluorescence
0.1 mg/mL
Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence
C20orf38, Chromosome 20 Open Reading Frame 38, dJ718P11, EC 2.3.1.50, hLCB2b, LCB 3, LCB2B, LCB3, Long Chain Base Biosynthesis Protein 2b, Long Chain Base Biosynthesis Protein 3, Serine Palmitoyltransferase 3, Serine Palmitoyltransferase, Long Chain Base Subunit 2-Like (Aminotransferase 2), Serine Palmitoyltransferase, Long Chain Base Subunit 3, Serine-Palmitoyl-CoA Transferase 3, SPT 3, SPT3, SPTLC2L
Rabbit
Affinity Purified
RUO
55304
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only