missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SPTLC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30881-25ug
This item is not returnable.
View return policy
Description
SPTLC1 Polyclonal antibody specifically detects SPTLC1 in Human samples. It is validated for Western Blot
Specifications
| SPTLC1 | |
| Polyclonal | |
| Western Blot : 1-5 μg/mL | |
| EC 2.3.1.50, hereditary sensory neuropathy, type 1, HSAN1, LBC1, LCB 1, LCB1HSAN, Long chain base biosynthesis protein 1, MGC14645, serine C-palmitoyltransferase, serine palmitoyltransferase 1, serine palmitoyltransferase, long chain base subunit 1, Serine-palmitoyl-CoA transferase 1, SPT 1, SPT1HSN1, SPTI | |
| Synthetic peptide derived from the human SPTLC1. | |
| 25 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 10558 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction