missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPTBN4 Antibody (S393-2), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59321FR
This item is not returnable.
View return policy
Description
SPTBN4 Monoclonal specifically detects SPTBN4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| SPTBN4 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SPTBN4 | |
| Fusion protein amino acids 1621-1832 (C-terminal repeats 14 to 15) of human Beta4-spectrin | |
| 100 μL | |
| 57731 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| S393-2 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| beta-IV spectrin, KIAA1642, QV, spectrin beta chain, brain 3, spectrin, beta, non-erythrocytic 4, spectrin, non-erythroid beta chain 3, SPNB4, SPTBN3 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Detects >200kDa. Does not cross react with other Beta-spectrins | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction