missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPTBN1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25681-1-AP-150UL
This item is not returnable.
View return policy
Description
Spectrin (Sp), the most abundant of the erythrocyte membrane skeleton proteins, helps these cells maintain their characteristic biconcave shape while remaining flexible and elastic. Erythrocyte Sp is a heterodimer composed of a 280 kDa alpha subunit and a 246 kDa beta subunit which associate in a side-to-side, antiparallel configuration to form a 100 nm rod-like structure. Sp in other tissues may be composed of distinct but homologous alpha and beta subunits, sometimes referred to as fodrin. A newly introduced nomenclature designates the Sp subunits of the erythrocyte as alpha-1 and beta-1, and the fodrin subunits as alpha-2 and beta-2. Alternatively spliced forms of each are designated as epsilon-1, epsilon-2, etc. (e. g. beta-1 epsilon-1).Specifications
| SPTBN1 | |
| Polyclonal | |
| Unconjugated | |
| SPTBN1 | |
| Beta II spectrin, betaSpII, ELF, Fodrin beta chain, Spectrin beta chain, brain 1, SPTB2, SPTBN1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20742, 6711 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q01082, Q62261 | |
| SPTBN1 | |
| SPTBN1 Fusion Protein Ag22459 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction