missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPTBN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552970
This item is not returnable.
View return policy
Description
Immunogen sequence: PLATSTDHGH NLQTVQLLIK KNQTLQKEIQ GHQPRIDDIF ERSQNIVTDS SSLSAEAIRQ RLADLKQLWG LLIEETEKRH RRLEEAHRAQ QYYFDAAEAE AWMSEQEL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
Spectrin (Sp), the most abundant of the erythrocyte membrane skeleton proteins, helps these cells maintain their characteristic biconcave shape while remaining flexible and elastic. Erythrocyte Sp is a heterodimer composed of a 280 kDa alpha subunit and a 246 kDa beta subunit which associate in a side-to-side, antiparallel configuration to form a 100 nm rod-like structure. Sp in other tissues may be composed of distinct but homologous alpha and beta subunits, sometimes referred to as fodrin. A newly introduced nomenclature designates the Sp subunits of the erythrocyte as alpha-1 and beta-1, and the fodrin subunits as alpha-2 and beta-2. Alternatively spliced forms of each are designated as epsilon-1, epsilon-2, etc. (e. g. beta-1 epsilon-1).
Specifications
| SPTBN1 | |
| Polyclonal | |
| Unconjugated | |
| SPTBN1 | |
| 9930031C03Rik; A2a; a--fodrin; AI842465; AL033301; alpha II spectrin; alpha-fodrin; alpha-II spectrin; alpha-spectrin 2; beta fodrin; beta II spectrin-like (short form); beta-fodrin; beta-G spectrin; beta-I spectrin; Beta-II spectrin; beta-spectrin 1; beta-spectrin 2; beta-spectrin 2, non-erythrocytic; beta-spectrin II; beta-spectrin non-erythrocytic 1; betaSpII; brain alpha-spectrin; brain erythroid spectrin (235E); brain spectrin; bSPII-; cytoskeleton protein; D330027P03Rik; EL3; ELF; elf1; elf3; embryonic liver beta-fodrin; embryonic liver fodrin; embryonic liver fodrin beta chain; epididymis luminal protein 102; erythroid spectrin beta; fodrin alpha chain; Fodrin beta chain; Gm1301; HEL102; HS2; HSPTB1; inhibitory protein factor; IPF; ja; jaundiced; membrane cytoskeletal protein; mKIAA4049; mKIAA4219; noerythroid alpha-spectrin 2; non-erythrocytic; nonerythroid alpha-spectrin 2; non-erythroid spectrin beta; short form of beta II spectrin; Sp beta; spectrin alpha chain, brain; spectrin alpha chain, non-erythrocytic 1; spectrin beta 1; spectrin beta 2; spectrin beta chain, brain 1; spectrin beta chain, erythrocyte; spectrin beta chain, erythrocytic; Spectrin beta chain, non-erythrocytic 1; spectrin beta Tandil; spectrin beta, erythrocytic; spectrin beta, non-erythrocytic 1; spectrin G; spectrin R; spectrin, alpha, non-erythrocytic 1; spectrin, beta, erythrocytic; spectrin, beta, non-erythrocytic 1; spectrin, non-erythroid alpha chain; spectrin, non-erythroid beta chain 1; SPH2; Spna2; Spnb1; Spnb-1; Spnb2; Spnb-2; Spta2; Sptan1; SPTB; SPTB1; Sptb2; Sptbn1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6711 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q01082 | |
| SPTBN1 | |
| Recombinant protein corresponding to Human Spectrin beta-1. Recombinant protein control fragment (Product #RP-89904). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction