missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spt6 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93722-0.1ml
This item is not returnable.
View return policy
Description
Spt6 Polyclonal antibody specifically detects Spt6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| Spt6 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunoprecipitation 1:100 - 1:500 | |
| emb-5, hSPT6, KIAA0162MGC87943, SPT6HSPT6, suppressor of Ty (S.cerevisiae) 6 homolog, suppressor of Ty 6 homolog (S. cerevisiae), Tat-cotransactivator 2 protein, Tat-CT2 protein, transcription elongation factor SPT6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1500-1726 of human SUPT6H (NP_003161.2). IFPTVNGLFRWFKDHYQDPVPGITPSSSSRTRTPASINATPANINLADLTRAVNALPQNMTSQMFSAIAAVTGQGQNPNATPAQWASSQYGYGGSGGGSSAYHVFPTPAQQPVATPLMTPSYSYTTPSQPITTPQYHQLQASTTPQSAQAQPQPSSSSRQRQQQPKSNSHAAIDWGKMAEQWLQEKEAERRKQKQRLTPRPSPSPMIESTPMSIAGDATPLLDEMDR | |
| 0.1 mL | |
| Signal Transduction | |
| 6830 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction