missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPT5 Recombinant Rabbit Monoclonal Antibody (23GB4220), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | SPT5 |
|---|---|
| Clone | 23GB4220 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089260
![]() |
Invitrogen™
MA553712 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088652
|
Invitrogen™
MA553713 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPT5 Recombinant Monoclonal antibody specifically detects SPT5 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| SPT5 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O00267, O55201 | |
| 20924, 308472, 6829 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4220 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AL033283; AL033321; AU019549; DRB sensitivity-inducing factor 160 kDa subunit; DRB sensitivity-inducing factor large subunit; DSIF large subunit; DSIF p160; etID10129.15; etID37101.15; fb37g01; fb95e05; fog; foggy; hSPT5; protein foggy; Spt5; SPT5 homolog, DSIF elongation factor subunit; SPT5H; suppressor of Ty 5; suppressor of Ty 5 homolog; suppressor of Ty 5 homolog (S. cerevisiae); Supt5; Supt5h; supt5h protein; Tat-cotransactivator 1 protein; Tat-CT1; Tat-CT1 protein; Transcription elongation factor SPT5; transcription elongation regulator FOGGY; wu:fb37g01; wu:fb95e05; wu:fc10f02 | |
| A synthesized peptide derived from human SUPT5H (770-825AA). | |
| Primary | |
| -20°C | |
| SUPT5H |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title