missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SPT5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17742624
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17742624 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17742624 Supplier Invitrogen™ Supplier No. PA5121226

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: U-87MG, HeLa, A-549, Mouse liver. The target is usually found in the following locations: Nucleus. Immunogen sequence: LQNLWRDQRE EELGEYYMKK YAKSSVGETV YGGSDELSDD ITQQQLLPGV KDPNLWTVKC KIGEERATAI SLMRKFIAYQ FTDTPLQIKS VVAPEHVKGY IYVEAYKQTH VKQAIEGVGN LRLGYWNQQM VPIKEMTDVL KVVKEVANLK PKSWVRLKRG IYKDDIAQVD YVEPSQNTIS LKMIPRIDYD RIKARMSLKD WFAKRKKFKR PPQRLFDAEK IRSLGGDVAS DGDFLIFEGN RYSRKGFLFK SFAMSAVITE GVKPTLSELE KFEDQPEGID LEVVTESTGK EREHNFQPGD N.

Component of the DRB sensitivity-inducing factor complex (DSIF complex), which regulates mRNA processing and transcription elongation by RNA polymerase II. DSIF positively regulates mRNA capping by stimulating the mRNA guanylyltransferase activity of RNGTT/CAP1A. DSIF also acts cooperatively with the negative elongation factor complex (NELF complex) to enhance transcriptional pausing at sites proximal to the promoter. Transcriptional pausing may facilitate the assembly of an elongation competent RNA polymerase II complex. DSIF and NELF promote pausing by inhibition of the transcription elongation factor TFIIS/S-II. TFIIS/S-II binds to RNA polymerase II at transcription pause sites and stimulates the weak intrinsic nuclease activity of the enzyme. Cleavage of blocked transcripts by RNA polymerase II promotes the resumption of transcription from the new 3' terminus and may allow repeated attempts at transcription through natural pause sites. DSIF can also positively regulate transcriptional elongation and is required for the efficient activation of transcriptional elongation by the HIV-1 nuclear transcriptional activator, Tat. DSIF acts to suppress transcriptional pausing in transcripts derived from the HIV-1 LTR and blocks premature release of HIV-1 transcripts at terminator sequences. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPT5
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 2.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SUPT5H
Gene Accession No. O00267, O55201
Gene Alias AL033283; AL033321; AU019549; DRB sensitivity-inducing factor 160 kDa subunit; DRB sensitivity-inducing factor large subunit; DSIF large subunit; DSIF p160; etID10129.15; etID37101.15; fb37g01; fb95e05; fog; foggy; hSPT5; protein foggy; Spt5; SPT5 homolog, DSIF elongation factor subunit; SPT5H; suppressor of Ty 5; suppressor of Ty 5 homolog; suppressor of Ty 5 homolog (S. cerevisiae); Supt5; Supt5h; supt5h protein; Tat-cotransactivator 1 protein; Tat-CT1; Tat-CT1 protein; Transcription elongation factor SPT5; transcription elongation regulator FOGGY; wu:fb37g01; wu:fb95e05; wu:fc10f02
Gene Symbols Supt5, SUPT5H
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 125-425 of human SUPT5H/SPT5 (NP_003160.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20924, 6829
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.