Learn More
SPT3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540315
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: MRKDKKKLRR LLKYMFIRDY KSKIVKGIDE DDLLEDKLSG SNNANKRQKI Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 86%; Rat: 93%
SPT3 Polyclonal antibody specifically detects SPT3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
SPT3 | |
Polyclonal | |
Unconjugated | |
SUPT3H | |
Cdc5-like protein; D9509.12; Positive regulator of Ty transcription; Protein SPT3; SPT3; SPT3 homolog, SAGA and STAGA complex component; SPT3L; SPT3-like protein; suppressor of Ty 3 homolog; suppressor of Ty 3 homolog (S. cerevisiae); SUPT3H; Transcription initiation protein SPT3 homolog; transcriptional regulator SPT3; YDR392W | |
Rabbit | |
Protein A | |
RUO | |
8464 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O75486 | |
SUPT3H | |
Synthetic peptide directed towards the N-terminal of human SUPT3H | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |